EasySat™ Bucketless Floor Mops, Contec®
Supplier: Contec
BLMOP BLVKMOP BLVKMOPIR TBLVKMOP TBLVKMOPIR BLMOPQC
33501-534EA 779.11 CAD
33501-534 33501-536 33501-538 33501-540 33501-542 CA76377-432
EasySat™ Bucketless Floor Mops, Contec®
Mops
Designed specifically for cleanroom use, this mop system completely eliminates the need for bucket or wringer.
  • Hydrophilic polyester/clean ester, laser cut sealed edge foam material
  • Translucent tank allows user to see solution
  • Fully autoclavable for critical areas
  • Easy to use
  • Compatible with a wide range of solvents
System includes a 1.5 l (52 oz.) translucent solution tank that can clean up to 5,000 square feet. Tank weighs only 3.2 kg (7 lbs.) when full. A push button flow control releases cleaning solution on demand. The angled, 55.9 cm (22") mopMore Product Information