PharMed® BPT Biocompatible Tubing, Saint-Gobain Life Sciences
Supplier: Saint Gobain Life Sciences
AY242038 AY242029 AY242046
63007-319PK 333.11 CAD
63007-319 63007-324 63007-345 63008-848 63008-850 87004-134 87004-154 87004-156 87004-160 CA87004-156 87004-152
PharMed® BPT Biocompatible Tubing, Saint-Gobain Life Sciences
Tubing Transfer Tubing
PharMed® BPT is designed to maintain fluid integrity during fluid transport. Transporting biocompatible fluids through a peristaltic pump limits the risk of fluid contact with any portion of the pump itself. PharMed® BPT tubing has been formulated to withstand the rigors of peristaltic pumping action while providing the biocompatible fluid surface required in sensitive applications.
  • It can be autoclaved repeatedly, and withstands repeated CIP and SIP cleaning and sterilization
  • PharMed® BPT outlasts silicone tubing in peristaltic pumps by up to 30 times,
More Product Information
Order Now
SPECIFICATIONS
Model 63007-319 63007-324 63007-345 63008-848 63008-850 87004-134 87004-154 87004-156 87004-160 CA87004-156 87004-152
Durometer hardness Shore A, 64 Shore A, 64 Shore A, 64 Shore A, 64 Shore A, 64 Shore A, 64 Shore A, 64 Shore A, 64 Shore A, 64 Shore A, 64 Shore A, 64
Environmentally Preferable
Max. pressure 43.0 psi 17.0 psi 10.0 psi 30.0 psi 13.0 psi 78.0 psi 9.0 psi 17.0 psi 11.0 psi 17.0 psi 11.0 psi
Max. vacuum 29.9" 29.9" 25.0" 29.9" 25.0" 29.9" 10.0" 29.9" 15.0" 29.9" 15.0"