VWR® Amber Latex Rubber Tubing
Supplier: VWR International
62995-723 62996-145 62996-178 62996-203 62996-350 62996-382 62996-418 62996-440 62996-473 62996-509 62996-531 62996-564 62996-597 62996-622 62996-655 62996-666 62996-688 62996-699 69400-000
62995-723PK 515.13 CAD
62995-723 62996-145 62996-178 62996-203 62996-350 62996-382 62996-418 62996-440 62996-473 62996-509 62996-531 62996-564 62996-597 62996-622 62996-655 62996-666 62996-688 62996-699 69400-000
VWR® Amber Latex Rubber Tubing
Tubing Transfer Tubing
High-quality rubber tubing recommended for all kinds of glass connections where highly elastic, clinging tubing is desired.
  • Durometer hardness: Shore A, 35
  • Tensile strength: 3500 psi
  • Translucent and durable
Amber latex rubber tubing is available in 50' lengths on reels or in boxes.
Ordering information: This product comes in three pieces.