VWR® Latex Rubber Tubing, Black
Supplier: VWR International
62995-608 62995-630 62995-663 62995-670 62995-680 62995-696 62995-721 62995-618
62995-608PK 130.8 CAD
62995-608 62995-630 62995-663 62995-670 62995-680 62995-696 62995-721 62995-618
VWR® Latex Rubber Tubing, Black
Tubing Transfer Tubing
Black latex rubber tubing withstands repeated sterilization and has superior tensile strength.
  • Durometer hardness: Shore A, 35
  • Tensile strength: 3500 psi
  • Packaged on 50' reels or in box packs
Black rubber tubing has maximum resistance to aging. Withstands repeated sterilization.
Order Now
SPECIFICATIONS
Environmentally Preferable