MegaTip™ Swab Mops, Micronova
Supplier: Micronova
MT-6IR MT-6 MT-11 MT-11IR
43300-972CS 360.94 CAD
43300-972 11211-984 76323-714 76323-716
MegaTip™ Swab Mops, Micronova
Mops
The MegaTip™ is a substantial cleaning swab that will easily reach to clean recesses, corners, and angles between walls, ceilings, and other hard to reach areas. When used with the SSU series handle, MegaTip™ Swab Mops may be used for applications where mop or wipe use is not practical.
  • Polyester/foam swab tip for cleaning 4×1.5" (10×4cm) areas
  • Stainless steel adapter
  • Available in smooth (NovaPoly) or textured (PolySorb) polyester formats
  • Compatible with alcohol and strong disinfectants
  • Available gamma irradiated for aseptic areas and equipment
The foam tip swab slips onto the MegaTip™ stainless steel frame and adapter. The gamma irradiated polyester covered swabs are ideal for aseptic cleanrooms.