VWR® Red Vacuum Rubber Tubing
Supplier: VWR International
62995-059 62995-062 62995-095 62995-120 62995-153 62995-335 62995-368 62995-390 62995-426
62995-059PK 557.29 CAD
62995-059 62995-062 62995-095 62995-120 62995-153 62995-335 62995-368 62995-390 62995-426
VWR® Red Vacuum Rubber Tubing
Tubing Vacuum/Pressure Tubing
Natural rubber tubing with extra-heavy wall especially recommended for vacuum work.
  • Durometer hardness: Shore A, 50
  • Tensile strength: 2000 to 2600 psi
Has high resilience and good tensile and tear strength.