HS 260 Reciprocating Shakers, IKA® Works
Supplier: IKA Works
0003066601 0003066701
33998-368EA 11861.63 CAD
33998-368 33998-372
HS 260 Reciprocating Shakers, IKA® Works
Shakers Reciprocating Shakers
These compact, flat shakers feature electronic adjustment of the speed and timer
The basic model has a timer with a range of 5 to 50 minutes. The control model features a timer with a range of 0 to 9 hours, 59 minutes, as well as a digital display to allow users to read the speed, timer function, and operating modes at a glance. Control model also features integrated end point positioning for automated robot-controlled sampling. All functions on the control model can be operated and documented with labworldsoft® software using the RS-232 interface.
The universal platform holds various types of vessels, and features variable clamping rolls that allow for universal adaptation. Universal platform surface dimensions: 32x32cm (121/2x121/2"). The flask clamp platform is for shaking flasks, Erlenmeyer flasks, and bottles with a round cross-section. Accommodates fifty-six 25mL flasks, twenty-three 50 or 100mL flasks, eleven 200 or 250mL flasks, or nine 500mL flasks. Flask clamps are available separately. Flask clamp platform surface dimensions: 33x33cm (13x13"). The dish platform is for smooth movement of cell cultures, nutrient media, Petri dishes, culture bottles, and vessels with a low center of gravity. Dish platform surface dimensions: 32x32cm (121/2x121/2"). The separating funnel platform is for shaking out, salting out, extracting, eluting, and enriching in pear-shaped separating funnels. Accommodates six 50mL funnels, five 100mL funnels, three 250mL funnels, and three 500mL funnels. The three included clamping rolls are height-adjustable for adaptation to different separating funnel sizes. Separating funnels are secured with five included O-rings. Separating funnel platform surface dimensions: 33.4x42.5cm (131/8x163/4").
A wide range of attachment combinations makes it possible to use almost all vessel sizes and shapes.