KIMAX® Capillary Tubing, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
80600-114 80600-3
89003-298CS 1974.56 CAD
89003-298 89003-300
KIMAX® Capillary Tubing, Kimble Chase
Tubing Transfer Tubing
Tubing is made of KG-33 borosilicate glass, which has a low coefficient of expansion and a high resistance to chemical attack
Ends are cut. Length: 121.9cm ±0.6 (48" ±1/4).
Certifications: Meets requirements for Type I Class A, ASTM E438.