ERGOMAT® Industry Floor Mats ERGOMAT®
Supplier: Ergomat
ERGOMAT®
EX0416 EX0204 EX0410 EX0314 EX0309 EX0310 EX0304 EX0404 EX0411 EX0207 EX0206 EX0308 EX0304 EX0413 EX0306 EX0213 EX0312 EX0213 EX0211 EX0215 EX0212 EX0407 EX0214 EX0303 EX0217 EX0307 EX0419 EX0310 EX0207 EX0209 EX0419 EX0209 EX0220 EX0318 EX0216 EX0219 EX0218 EX0317 EX0217 EX0203 EX0208 EX0409 EX0316 EX0315 EX0406 EX0306 EX0313 EX0308 EX0315 EX0406 EX0318 EX0415 EX0320 EX0211 EX0316 EX0408 EX0311 EX0214 EX0420 EX0420 EX0320 EX0418 EX0210 EX0206 EX0405 EX0412 EX0219 EX0216 EX0203 EX0317 EX0413 EX0313 EX0409 EX0417 EX0303 EX0414 EX0408 EX0311 EX0305 EX0314 EX0208 EX0212 EX0305 EX0210 EX0411 EX0410 EX0418 EX0319 EX0220 EX0309 EX0218 EX0417 EX0404 EX0416 EX0415 EX0312 EX0319 EX0205 EX0204 EX0405 EX0205 EX0407 EX0215 EX0307 EX0412 EX0414
CA89170-030EA 1648.75 CAD
CA89170-030 89170-020 CA89170-032 89170-022 89170-024 CA89170-034 89170-026 CA89170-036 89170-028 CA89170-038 89170-030 89170-032 CA89170-040 89170-034 CA89170-042 CA89170-044 89170-036 CA89170-046 89170-038 CA89170-048 89170-080 CA89170-090 CA89170-092 89170-082 89170-084 CA89170-094 CA22231-188 CA89170-096 89170-086 CA22231-186 CA89170-098 CA89170-010 89170-088 CA89170-012 CA89170-014 89170-006 CA89170-016 CA89170-018 89170-008 89170-090 89170-092 89170-094 89170-096 22231-188 89170-010 89170-098 22231-186 CA89170-020 89170-012 CA89170-022 89170-014 CA89170-024 89170-016 CA89170-026 89170-018 CA89170-028 89170-060 CA89170-070 89170-062 CA89170-072 89170-064 CA89170-074 CA89170-076 89170-066 89170-100 89170-068 CA89170-078 89170-102 89170-104 89170-106 89170-070 89170-072 CA89170-080 89170-074 CA89170-082 CA89170-084 89170-076 CA89170-086 89170-078 CA89170-088 CA89170-006 CA89170-008 CA89170-050 89170-040 CA89170-052 89170-042 89170-044 CA89170-054 CA89170-056 89170-046 89170-048 CA89170-058 89170-050 89170-052 CA89170-060 CA89170-062 89170-054 89170-056 CA89170-064 CA89170-100 CA89170-066 89170-058 CA89170-102 CA89170-068 CA89170-104 CA89170-106
ERGOMAT® Industry Floor Mats ERGOMAT®
Floor Mats Traction and Anti-Fatigue Floor Mats
This heavy duty nitrile drainage mats is ideal for areas with extreme moisture or heavy fluid flow, from water to cutting oils
  • Heavy-duty nitrile
  • For areas with extreme moisture or heavy fluid flow, from water to cutting oils
  • Anti-slip surface and 5.1 cm (2") molded bevels for increased safety
  • Suitable for use in ISO Class 5 (FED-STD-209D Class 100/M3.5) cleanrooms
They offer an exceptional anti-slip surface and 5.1 cm (2") molded bevels for increased safety. Mats are typically used in metalwork, pulp and paper manufacturing, dairy farms, food and beverage industries, and car washes. Mats are suitable for use in ISO Class 5 (FED-STD-209D Class 100/M3.5) cleanrooms.Black color.
Ordering information: Manufacturer's five year warranty.