Spectra/Por® Weighted Closures, Spectrum® Laboratories
Supplier: Spectrum Laboratories
132742 132744 132746
25224-136PK 347.98 CAD
25224-136 25224-150 25224-154
Spectra/Por® Weighted Closures, Spectrum® Laboratories
Closures Test/Sample Tube Closures
Pressure-fitted with an acid-washed, stainless steel bar into the reverse side
Made of polypropylene. Autoclavable. When used in tandem with the standard closure, weighted closure facilitates complete submersion of the dialysis sac for a more efficient dialysis effect.