Spectra/Por® Magnetic Weighted Closures, Spectrum® Laboratories
Supplier: Spectrum Laboratories
132760 132762 132764 132766
25224-168PK 233.89 CAD
25224-168 25224-170 25224-172 25224-174
Spectra/Por® Magnetic Weighted Closures, Spectrum® Laboratories
Closures Test/Sample Tube Closures
Closures feature imbedded fluoropolymer resin-coated magnetic weights
Sink in water. Provide a slow spin movement of dialysis membrane when used with a magnetic stir plate. No additional stirring bar is required to maintain continuous motion. Not for use with cellulose ester (CE) or polyvinylidene fluoride (type F) tubing.
Order Now
SPECIFICATIONS
Environmentally Preferable