Spectra/Por® Universal Closures, Spectrum® Laboratories
Supplier: Spectrum Laboratories
142110 142250 142252 142113 142170 142172 142173 142174 142175 142176 142150 142152 142153 142154 142155 142253 142112 142156
25224-080PK 331.97 CAD
25224-080 25224-082 25225-292 25225-298 25225-300 25225-302 25225-304 25225-306 25225-308 25225-310 25225-312 25225-314 25225-316 25225-318 25225-320 25225-294 25225-296 25225-322
Spectra/Por® Universal Closures, Spectrum® Laboratories
Closures Test/Sample Tube Closures
Nylon closures lock gently over rigid membranes such as cellulose ester (CE) and polyvinylidene fluoride (type F) tubing, providing a strong, leakproof seal
Closures can also be used with traditional regenerated cellulose (RC) tubing. Sink in water. Can be sterilized with gamma radiation, ethylene oxide, ethanol, or formaldehyde. Not autoclavable. For best results, select a closure 4 to 10mm wider than the flat width of the membrane or tubing.
Order Now
SPECIFICATIONS
Environmentally Preferable