Tissue Marking Dyes (TMD™), TBS®, General Data
Supplier: General Data Healthcare
TMD-BK
TMD-Y
TMD-O
TMD-BK
TMD-BL
TMD-Y
TMD-R
TMD-G
15147-923CS
782.46
CAD
15147-923
15147-927
CA15148-232
CA27900-258
CA27900-260
CA27900-262
CA27900-264
CA27900-266
Tissue Marking Dyes (TMD™), TBS®, General Data
Stains
Histology Stains
Ideal for marking margins or for processing multiple specimens in one tissue cassette, these dyes have been reformulated for superior adherence to tissue surfaces
Viscosity allows a thin, even coating of pigment without masking underlying tissue. Quick drying decreases frozen sectioning time and provides excellent readability. Size: 237 mL (8 oz.).