HydroGOLD Capillary Columns
Supplier: Thermo Scientific
26H50-0232 26H50-0253 26H50-0525 26H50-0532 26H50-0553 26H50-1032 26H50-1053
10811-884EA 151.39 CAD
10811-884 10811-886 10811-890 10811-892 10811-894 10811-900 10811-902
HydroGOLD Capillary Columns
Chromatography Columns
Capillary HydroGOLD columns provide protection to the analytical column.
  • Focuses target analytes at the head of the analytical column, leading to better chromatographic peak shape
  • Highly deactivated to provide superior inertness, essential for analysis of active compounds
  • HydroGOLD extends column lifetime by preventing degradation from harsh 'steam-cleaning' water injections
Protects against column contamination caused by non-volatile materials, extending the column lifetime.