Automatic Retractable Cutters and Knife, Martor USA
Supplier: Martor
07152 08152
61200-308EA 47.76 CAD
61200-308 61200-312
Automatic Retractable Cutters and Knife, Martor USA
Knives
Optisafe and Maxisafe knives feature a safety spring that allows the automatic blade to “shoot-back” into the handle even in response to any uncontrolled movement
The Multisafe safety knife has a die-cast zinc body and spring-loaded blade slider on top for horizontal cutting work only.
The Profi automatic retractable safety knife is equipped with a lightweight aluminum body and a deep-edged trapezoid blade. The spring-loaded blade retract system is designed to respond to any uncontrolled movements, or when the thumb is removed from the slider.
The Ultrasafe safety knife with automatic open-frame blade guarding function is ideal for safe opening of plastic and paper sacks.
The Maxisafe knife has a longer blade extension than the Optisafe knife. The Maxisafe three-sided cutter is similar to the Maxisafe knife, except it features a three-sided blade slider.