TraceGOLD™ TG-624SiLMS GC Columns
Supplier: Thermo Scientific
26059-3390 26059-3410 26059-3960 26059-4950 26059-3320 26059-3330 26059-4080
10056-644EA 1765.25 CAD
10056-644 10056-646 10056-648 10056-816 10056-818 10056-820 10057-138
TraceGOLD™ TG-624SiLMS GC Columns
Chromatography Columns
Analyze residual solvents, volatile organic compounds, alcohols, and oxygenates using Thermo Scientific™ TraceGOLD™ TG-624SilMS GC Columns. Both phases offer the low to mid polarity phase, 6% cyanopropylphenyl methypolysilozane.
TraceGOLD TG-624SilMS GC Columns provide high thermal stability with maximum temperatures up to 320 °C, which is ideal for volatile organics analysis. They are highly inert, providing excellent peak shape for a wide range of compounds.
Applications includes Residual solvents, volatile organic compounds, alcohols, oxygenates.