ProPac™ Elite WCX HPLC Columns
Supplier: Thermo Scientific
303028 303027 303026 302973 302972 302976 302977 303355
76276-276EA 2159.5 CAD
76276-276 76276-278 76276-280 76276-282 76276-284 76276-288 76276-290 76477-166
ProPac™ Elite WCX HPLC Columns
Chromatography Columns
Thermo Scientific™ ProPac™ Elite WCX weak cation exchange (WCX) liquid chromatography column.
  • Superior resolution power for proteins, monoclonal antibodies, and associated charge variants
  • High efficiency with reproducible separations
  • High recovery with low carry-over
  • Wide pH operating range: 2 to 12
  • High temperature stability: up to 60 °C
  • High throughput
  • Compatible with CX-1 pH gradient buffers
Designed for protein characterization including therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs) and biosimilar proteins,More Product Information