Rotary Evaporators, RV 8
Supplier: IKA Works
0010003571 0010003572 0010003570
76329-690EA 9769.28 CAD
76329-690 76329-884 76329-908
Rotary Evaporators, RV 8
Evaporators Rotary Evaporators
These entry-level evaporators are ideal for all standard evaporation applications in universities and research laboratories. The functions of the RV10 control have been extended and improved to enable fully automatic distillation: With automatic boiling point determination, an integrated solvent library and the option to connect a speed-controlled vacuum pump.
  • All models have manual lift with safety stop function
  • Digital TFT display showing the rotation speed and heating bath temperature allows optimal control of the entire distillation process
  • Single-hand lift system with pressure sensors can be operated easily with the right or left hand
  • Water/oil heating bath can be used as a stand-alone unit and has integrated ergonomic carrying handles
  • New clamp mechanism for an easier changing of evaporation flasks
  • USB connection enables online update of the firmware
Delivery information: Units are supplied with heating bath HB 10, either with or without glassware. RV 8 models are compatible with the full RV10 glassware range.
Please note: A vacuum source, vacuum check valve and cooling system are required but not supplied. Please contact your local Avantor sales office for advice and details.
Caution: * Flex package is delivered with bath, vapour tube, clamps for glassware and Woulf bottle only. This package enables the customer to select the glassware set up for their specific application.