Baths for Immersion Thermostats, Plastic, IB R eco
Supplier: IKA Works
0020004383 0020004382
76499-890EA 1721.63 CAD
76499-890 76499-900
Baths for Immersion Thermostats, Plastic, IB R eco
Baths Water Baths, Unstirred
Rectangular plastic baths for use with immersion thermostats.
Max. temperature: 100 °C; water only