TraceGOLD TG-ALC Plus ll Columns
Supplier: Thermo Scientific
26063-2240 26063-2980
10057-160EA 1769.47 CAD
10057-160 10057-162
TraceGOLD TG-ALC Plus ll Columns
Chromatography Columns
Application-specific columns for blood alcohol analysis.
  • Blood alcohol analysis
  • Abused inhalent anesthetics
  • Common industrial solvents
Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity. Fast analysis time giving full separation of blood alcohols