TraceGOLD TG-ALC Plus l Columns
Supplier: Thermo Scientific
26063-3390 26063-3960
10057-156EA 1753.41 CAD
10057-156 10057-158
TraceGOLD TG-ALC Plus l Columns
Chromatography Columns
Application-specific columns for blood alcohol analysis.
  • Blood alcohol analysis
  • Abused inhalent anesthetics
  • Common industrial solvents
Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity. Fast analysis time giving full separation of blood alcohols