Guard Column Holders for Preparative HPLC Columns
Supplier: Thermo Scientific
885-010 887-020 887-030 887-031 891-030 891-031
76332-084EA 494.73 CAD
76332-084 76332-126 76332-128 76332-130 76332-134 76332-136
Guard Column Holders for Preparative HPLC Columns
Guard Column Holders
Using preparative guards is a proven and cost effective way to extend the lifetime of your preparative columns. Thermo Scientific™ Preparative Guard Column Holders allow for easy assembly and connection of the guard column to the preparative column.
A range of guard holder sizes are offered, which are all in a convenient stand-alone format that requires connections from the LC inlet and outlet to the preparative LC column.
Preparative guard holders are available in ¹/₁₆ or ¹/₈" connection fittings in the larger guard sizes to be compatible with your Preparative HPLC system hardware.