Tissue Capsules, Simport
Supplier: Simport Scientific
M471 M470
CA73960-012CS 487.45 CAD
CA73960-012 CA73960-010
Tissue Capsules, Simport
Embedding Cassettes Tissue Capsules
These polypropylene capsules are suitable for holding tissue samples during processing
The lids have a frosted marking area for sample identification and an open mesh area to facilitate fluid exchange. The whole surface of the base is also an open mesh. The lid snaps securely on the base, eliminating the risk of tissue loss during processing. Color: white.