SureSTART™ WebSeal™ 96-Well Plate Sealing Mats for Well Plate Kits
Supplier: Thermo Scientific
60180-M152 60180-M153 60180-M154 60180-M151
10036-046PK 415.15 CAD
10036-046 10036-048 10036-050 10036-308
SureSTART™ WebSeal™ 96-Well Plate Sealing Mats for Well Plate Kits
Seals Microplate Seals
Save time and minimize the risk of cross-contamination with Thermo Scientific™ SureSTART™ Level 3 WebSeal™ 96-well plate sealing mats for well plate kits. Select SureSTART Level 3 products for ultimate performance and sensitivity from your methods while maintaining the highest levels of sample security and integrity.
  • Ideal for regulated labs using all solvent types including non-polar solvents
  • Suitable for HPLC/UHPLC, QA/QC, and sensitive applications
  • Ideal for verifying compound presence and quantification using detection techniques such as UV, DAD/PDA, RI, fluorescence, and ELSD
  • Eliminate cross contamination of samples
  • Resist coring and tearing
  • Superior resealability after multiple injections
  • WebSeal mats are manufactured from EVA (Ethylene-vinyl acetate) or pure silicone with or without a PTFE coating
  • Silicone mats are dry heat autoclavable with excellent chemical compatibility, and can withstand low temperatures down to −80 °C
  • 100% traceable through the manufacturing process
  • Certificate of Conformance available by lot number upon request
Level 3 products are for high performance applications and are suitable for regulated labs using all solvent types including non-polar solvents.
Silicone mats - Using a silicone mat with the correct profile reduces sample and solvent evaporation during your workflows, provides better consistency, and is ideal for long-term storage.
Certifications: These 96-well plate sealing mats are specification certified to ensure autosampler compatible physical dimensions and conform to ANSI-SLAS 4-2004 standards.