Swingsette™ M517 Tissue Processing/Embedding Cassettes, Simport
Supplier: Simport Scientific
M517-3 M517-4 M517-5 M517-6 M517-7 M517-8 M517-9 M517-11 M517-12 M517-16 M517-18 M517-2 M517-10
CA97000-504CS 261.76 CAD
CA97000-504 CA97000-506 CA97000-508 CA97000-510 CA97000-512 CA97000-514 CA97000-516 CA97000-520 CA97000-522 CA97000-524 CA97000-528 CA97000-502 CA97000-518
Swingsette™ M517 Tissue Processing/Embedding Cassettes, Simport
Embedding Cassettes Tissue Processing Cassettes
Disposable plastic cassettes hold tissue specimens during processing, embedding, and storage
A special hinge holds the base and lid together and allows cassettes to be opened and closed as often as necessary. The lid can be removed and reinserted easily without the danger of specimen loss. Anterior surface is slanted at a 45° angle for use with leading labeling instruments. Three frosted writing surfaces. Cassette colors are non-cytotoxic and non-metallic.
Most convenient for cassette labeling instruments, as the lids are packaged separately in the case. They are molded from a special,More Product Information