VWR® Thin Sleeves, PTFE
Supplier: VWR
VWR®
019.024 019.019.1 019.029 019.014.1 019.050 019.045
89026-484PK 184.62 CAD
89026-484 89026-496 89026-480 89026-490 89026-488 89026-498
VWR® Thin Sleeves, PTFE
NS Joint Sleeves and Connectors Joint Sleeves
These PTFE sleeves withstand low to medium pressure and are inserted between standard ground glass joints to eliminate the use of grease and prevent sticking.
  • Chemically resistant
  • Inert
  • PTFE fabrication
Sleeves are inert, have a wall thickness of about 0.05mm, and are usable to 280°C (536°F).