Phlebotomy Tray Units, JAC Medical
Supplier: JAC
JAC8 JAC9 JAC10 JAC58 JAC59 JAC60
CA78010-016EA 178.41 CAD
CA78010-016 CA78010-018 CA78010-020 CA78010-144 CA78010-146 CA78010-148
Phlebotomy Tray Units, JAC Medical
Trays/Dishes/Pots Blood Collection Trays
Polystyrene trays include components to assist in efficient blood collection
All replacement components (except HDPE containers) are constructed of translucent white autoclavable polypropylene. The utility boxes contain one removable divider and are useful for carrying a variety of small items. The gauze box has one removable divider that creates three compartments total: one large 11.7Wx11.7Dcm (45/8x45/8") compartment and two smaller 6.4Wx7Dcm (21/2x23/4") compartments. The packet/tube holder is ideal for BD Vacutainer® tubes, packets, winged infusion sets, and syringes. It features three removable dividers to provide up to six compartments. The 237mL (8oz.) HDPE containers feature snap-on lids and have excellent resistance to acids, alcohols, and bases.
Ordering information: Mini tray unit 30109-522 is supplied with one 11-well test tube rack, two mini utility boxes, one small waste holder, and two 237mL (8oz.) HDPE containers with lids. Medium tray units 30109-520 and 62687-300 are supplied with a test tube rack, two utility boxes, one small waste holder, two 237mL (8oz.) HDPE containers with lids, one slip holder, and one slide holder.