HISTOSETTE® II Biopsy Processing and Embedding Cassettes, Simport Scientific
Supplier: Simport Scientific
M486-2 M486-3 M486-4 M486-5 M486-6 M486-7 M486-8 M486-9 M486-10 M486-11 M486-12
CA87002-346CS 356.39 CAD
CA87002-346 CA87002-348 CA87002-350 CA87002-352 CA87002-354 CA87002-356 CA87002-358 CA87002-360 CA87002-362 CA87002-364 CA87002-366
HISTOSETTE® II Biopsy Processing and Embedding Cassettes, Simport Scientific
Embedding Cassettes Biopsy Processing Cassettes
These disposable plastic cassettes hold tissue or biopsy specimens during processing and embedding as well as in storage
Made of a high-density acetal polymer, the cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. The efficient 1mm square openings maximize fluid exchange and ensure proper drainage. Large labeling areas are located on three sides of the cassettes. The anterior writing area is slanted at a 45° angle to make the cassette more compatible with cassette labeling instruments. Colors are non-cytotoxic and nonmetallic.
Ordering information: Lids are supplied in unattached. Packed in two dispenser boxes of 500 cassettes and one dispenser box of 1000 lids.