VWR® Mortars and Pestles, Porcelain
Supplier: VWR International
89038-158 89038-174 89038-144 89038-146 89038-148 89038-150 89038-152 89038-154 89038-156 89038-160 89038-162 89038-164 89038-166 89038-168 89038-170 89038-172 89038-176 89038-178 89038-180 89038-182 89038-184
89038-158CS 658.15 CAD
89038-158 89038-174 89038-144 89038-146 89038-148 89038-150 89038-152 89038-154 89038-156 89038-160 89038-162 89038-164 89038-166 89038-168 89038-170 89038-172 89038-176 89038-178 89038-180 89038-182 89038-184
VWR® Mortars and Pestles, Porcelain
Mortars and Pestles Mortars with Pestles
Mortars are constructed of chemical porcelain and feature a lip and a glazed exterior.
  • Porcelain construction
  • Glazed exterior, unglazed interior
  • Oversized pestle for comfort
Pestles are constructed entirely of chemical porcelain or with a porcelain grinding head and wooden handle. Wooden handle is not autoclavable. Pestles are also glazed to grinding surface and oversized for hand comfort.