Embedding Rings, Simport Scientific
Supplier: Simport Scientific
M460 M460-4 M460-6 M460-3 M460-5
CA48648-000CS 439.65 CAD
CA48648-000 CA73960-004 CA73960-008 CA73960-002 CA73960-006
Embedding Rings, Simport Scientific
Embedding Cassettes Embedding Rings
Made of high-impact polystyrene, these embedding rings are suitable for holding and identifying tissue sample blocks
The rings fit well in microtome chuck adapters. The etched writing surface on the ring is marked with an identification number and placed on top of the sample block. Additional paraffin is poured into the base mold to cement the ring onto the tissue block. The embedding ring securely holds the tissue sample in the microtome chuck adapter for sectioning and then identifies the sample while in storage.
Ordering information: Each case contains four dispenser boxes of 250 rings.