HISTOSETTE® II Cassettes in E-Z LOAD™ Sleeves, Simport
Supplier: Simport Scientific
M482-2BA M482-3BA M482-4BA M482-5BA M482-6BA M482-7BA M482-8BA M482-9BA M482-10BA M482-12BA M482-11BA M482-2LI M482-3LI M482-4LI M482-5LI M482-6LI M482-7LI M482-8LI M482-9LI M482-10LI M482-11LI M482-12LI M483-2BA M483-3BA M483-4BA M483-5BA M483-6BA M483-7BA M483-8BA M483-9BA M483-10BA M483-12BA M483-2LI M483-3LI M483-4LI M483-5LI M483-6LI M483-7LI M483-8LI M483-9LI M483-10LI M483-11LI M483-12LI
CA60830-008CS 313.46 CAD
CA60830-008 CA60830-010 CA60830-012 CA60830-014 CA60830-016 CA60830-018 CA60830-020 CA60830-022 CA60830-024 CA60830-028 CA60830-029 CA60830-030 CA60830-032 CA60830-034 CA60830-036 CA60830-038 CA60830-040 CA60830-042 CA60830-044 CA60830-045 CA60830-048 CA60830-050 CA60830-052 CA60830-054 CA60830-056 CA60830-058 CA60830-060 CA60830-062 CA60830-064 CA60830-066 CA60830-068 CA60830-072 CA60830-074 CA60830-076 CA60830-078 CA60830-080 CA60830-081 CA60830-084 CA60830-086 CA60830-088 CA60830-090 CA60830-092 CA60830-094
HISTOSETTE® II Cassettes in E-Z LOAD™ Sleeves, Simport
Embedding Cassettes Cassettes for Labeling Instruments
Molded from a high-density acetal polymer, these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents
CE marked.
The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The anterior printing area is at a 45° angle, offering an unobstructed view of the writing surface and making the cassettes compatible with automated labeling instruments. Biopsy cassettes are designed to hold biopsy samples securely during the embedding process. One-piece disposable plastic lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss.
Ordering information: Cassette bases are packaged in E-Z LOAD™ sleeves of 50 bases, secured by a holding tie. Each case contains two boxes of 20 sleeves.