Slimsette™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
Supplier: Simport Scientific
M509-2SL M509-3SL M509-4SL M509-5SL M509-6SL M509-7SL M509-8SL M509-9SL M509-10SL M509-11SL M509-12SL M509-16SL M509-17SL M509-18SL M510-2SL M510-3SL M510-4SL M510-5SL M510-6SL M510-7SL M510-8SL M510-9SL M510-10SL M510-11SL M510-12SL M510-16SL M510-17SL M510-18SL
CA95030-230CS 597.49 CAD
CA95030-230 CA95030-232 CA95030-234 CA95030-236 CA95030-238 CA95030-240 CA95030-242 CA95030-244 CA95030-246 CA95030-248 CA95030-250 CA95030-252 CA95030-254 CA95030-256 CA95030-258 CA95030-260 CA95030-262 CA95030-264 CA95030-266 CA95030-268 CA95030-270 CA95030-272 CA95030-274 CA95030-276 CA95030-278 CA95030-280 CA95030-282 CA95030-284
Slimsette™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
Embedding Cassettes Cassettes for Labeling Instruments
These cassettes are made of a high-density acetal polymer and keep specimens safely submerged in liquid
The cassettes are totally resistant to the chemical action of histological solvents. The flow-through openings allow for maximum fluid exchange and ensure proper drainage. The compact design features a space-saving recessed cover. The one-piece integral lid eliminates the need for separate steel lids. Lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassettes. TheMore Product Information