UVP Converter Plates, Analytik Jena
Supplier: Analytik Jena CA
38-0191-02 38-0200-01 38-0191-01 38-0191-04
95025-406EA 1493.38 CAD
95025-406 CA11500-064 CA15000-088 CA21474-892 CA82026-878
UVP Converter Plates, Analytik Jena
Transilluminators UV/White Light Transilluminators
UV to White Light Converter Plate measuring 21 x 26 cm, converts Analytik Jena transilluminator’s ultraviolet light to white light for viewing protein gels and Coomassie blue stained or silver stained media
Visi-Blue™ converter plate is designed to convert 302 nm UV light to 480 nm blue light for use with SYBR® Green, SYPRO® Orange and GFP stains. Surface is safety-fused into metal frame for durability.
Phosphored plexiglass uses a white diffuser to convert UV radiation. Plate housing is scratch-resistant metal. Handles on two sides allow easy transport.