SWINGSETTE™ M518 Biopsy Processing and Embedding Cassettes, Simport Scientific
Supplier: Simport Scientific
M518-2 M518-3 M518-4 M518-5 M518-6 M518-7 M518-8 M518-9 M518-10 M518-11 M518-12 M518-16 M518-17 M518-18
CA97000-530CS 249.03 CAD
CA97000-530 CA97000-532 CA97000-534 CA97000-536 CA97000-538 CA97000-540 CA97000-542 CA97000-544 CA97000-546 CA97000-548 CA97000-550 CA97000-552 CA97000-554 CA97000-556
SWINGSETTE™ M518 Biopsy Processing and Embedding Cassettes, Simport Scientific
Embedding Cassettes Biopsy Processing Cassettes
These disposable plastic cassettes hold biopsy specimens during processing, embedding, and storage
A special hinge holds the base and lid together and allows cassettes to be opened and closed as often as necessary. The lid can be removed and reinserted easily without the danger of specimen loss. Anterior surface is slanted at a 45° angle for use with leading labeling instruments. Three frosted writing surfaces. Cassette colors are non-cytotoxic and non-metallic.
Most convenient for cassette labeling instruments, as the lids are packaged separately in the case. They are molded from aMore Product Information