SWINGSETTE™ M516 Biopsy Processing and Embedding Cassettes, Simport Scientific
Supplier: Simport Scientific
M516-2 M516-3 M516-4 M516-5 M516-6 M516-7 M516-8 M516-9 M516-10 M516-11 M516-12 M516-16 M516-17 M516-18
CA97000-474CS 482.03 CAD
CA97000-474 CA97000-476 CA97000-478 CA97000-480 CA97000-482 CA97000-484 CA97000-486 CA97000-488 CA97000-490 CA97000-492 CA97000-494 CA97000-496 CA97000-498 CA97000-500
SWINGSETTE™ M516 Biopsy Processing and Embedding Cassettes, Simport Scientific
Embedding Cassettes Biopsy Processing Cassettes
These disposable plastic cassettes are specially designed to hold biopsy specimens during processing, embedding, and storage
A special hinge holds the base and cover together and allows cassettes to be opened and closed as often as necessary. The cover can be removed and reinserted easily without the danger of specimen loss. Anterior surface is slanted at a 45° angle for use with leading labeling instruments. Three frosted writing surfaces. Cassette colors are non-cytotoxic and non-metallic.
They are molded from a special, high-density acetal polymer and are completely resistant to theMore Product Information