MICROSETTE™ Biopsy Cassettes with 6 Compartments, Simport
Supplier: Simport Scientific
M503-2 M503-3 M503-5 M503-7 M503-9 M503-10 M503-11 M503-9SL
CA48648-300CS 391.43 CAD
CA48648-300 CA48648-301 CA48648-303 CA48648-304 CA48648-305 CA48648-306 CA48648-307 CA48648-308 CA48648-309 CA73960-543 CA73960-545 CA73960-547 89511-286 89511-288 89511-290 CA76642-510
MICROSETTE™ Biopsy Cassettes with 6 Compartments, Simport
Embedding Cassettes Biopsy Processing Cassettes
Molded from acetal, these patented cassettes keep specimens safely submerged in liquid and are resistant to most histological solvents. The anterior writing area is at a 45° angle to make the cassette more suitable to be used with most types of cassette labeling instruments.
  • With six separate compartments
  • Unobstructed anterior writing area at a 45° angle
  • No biopsy pad necessary
  • Compatible with most cassette printers
  • Lid is pre-attached to base when bulk
  • Lid is seperate when in sleeve
This model can hold up to six tissue specimens, each one placed in its own 7×12 mm (¹/₄×⁷/₁₆") compartment, numbered from 1 to 6. Cover and base have over 2000 square openings to maximize fluid exchange and ensure proper drainage. Approximately 170 holes (each having a diameter of 0.26 mm) per compartment.
Packaging: The bulk cassettes contain four dispenser boxes of 250 cassettes per case. The sleeved cassette model has 10 sleeves of 75 cassettes per case.