Sanitizing Footbath Mats, Wearwell®
Supplier: Wearwell
222.12X24X32BK 560.78X6X18BK-CS10
89021-490EA 214.5 CAD
89021-490 89025-906
Sanitizing Footbath Mats, Wearwell®
Floor Mats Tacky Mats
Mats keep hygienic areas free of contamination and germs
  • Keeps hygienic areas free of contamination and germs
  • Thousands of pliable rubber scrapers dislodge contaminants from footwear as soles are immersed in disinfectant solution
  • Ideal for food-processing plants
The 61x81.3cm (24x32") mat is 1.3cm (1/2") thick and holds 3.8L (1gal.) of chlorine or quaternary disinfectant solution. The 81.3x99cm (32x39") tall-wall mat has 6.4cm (21/2") sides that allow it to hold up to 20.8L (5.5gal.) of sanitizing solution. When used to fill the mat, the solution will totally immerse the treads and sides of boot soles, while the fingertips release any accumulations.
Ideal for food processing plants, these mats have thousands of pliable rubber scrapers that dislodge contaminants from footwear as the shoe soles are immersed in the disinfectant solution.