24/Seven™ Modular Floor Mats, Wearwell®
Supplier: Wearwell
570.58X3X3GRBK 572.EDGINGFGRYL 572.EDGINGMGRYL 570.58X3X3CFRBK 572.EDGINGFCFRYL 572.EDGINGMCFRYL 570.58X3X3NBRBK 572.58X3X3NBRBK 572.EDGINGMNBRYL 572.58X3X3GRBK
89021-144EA 151.24 CAD
89021-144 89021-222 89021-226 89021-362 89021-372 89021-382 89021-384 89040-392 89040-394 89040-402 89040-404 89040-406 89021-146
24/Seven™ Modular Floor Mats, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
These 0.9x0.9m (3x3') modular, molded rubber antifatigue mats are resilient and comfortable.The mats are available with a functional and effective drainage surface, or with a solid surface for use in mostly dry areas. Both styles are available in three unique compounds to address the diverse environments found in typical work applications.
  • The grease-resistant mats are ideal for general-purpose applications
  • The cutting-fluid-resistant (CFR) compound mats are ideal for areas with mineral oil-based cutting fluids
  • The 100% nitrile compound mats are ideal for areas with petroleum-based fluids, such as hydrocarbons, naphthenes, paraffins, petroleum distillates, and other petrochemicals
  • For additional traction GritWorks!® surface on the CFR or nitrile mats: provides up to 40% more traction than recommended by OSHA in wet areas
  • Single workstation mats also have a hidden interlocking system designed to connect on all four sides to quickly cover large areas
For extra safety, attach optional yellow 7.6Wx99Lcm (3x39") beveled ramp edges. Female ramps snap onto the protruding mat knobs; male ramps snap into the underside of the mat.
All mats are 1.6 cm (⅝") thick.