Nalgene® 870 PFA Tubing, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
8051-0125 8051-0375 8051-0187 8051-0250
63014-260CS 712.34 CAD
63014-260 63014-262 63014-261 63014-264
Nalgene® 870 PFA Tubing, Thermo Scientific
Tubing Transfer Tubing
For low-pressure use with mechanical, compression-type fittings.
  • PFA tubing is translucent and semi-rigid with very high chemical resistance
  • Very low water absorption
Wide temperature range: –270 to +260 °C (–454 to +500 °F). Durometer hardness: Shore D, 55.
Can be sterilized by autoclaving, gas, dry heat, and disinfectants. Do not sterilize by radiation. Resin complies with USDA regulations.
Ordering information: Supplied in 25' coil lengths.