VYCOR® Glass Tubing, Standard Wall, ProScience Glass
Supplier: Proscience
GS32835-023 GS32835-045 GS32835-067 GS32835-205 GS32835-249 GS32835-409
CA87003-582EA 40.67 CAD
CA87003-582 CA87003-584 CA87003-586 CA87003-588 CA87003-590 CA87003-592
VYCOR® Glass Tubing, Standard Wall, ProScience Glass
Tubing Transfer Tubing
Well-suited for precise analytical work
With a high thermal shock resistance and a low coefficient of expansion, VYCOR® 96% pure silica glass withstands temperatures up to 900°C (1652°F). Softening point is approximately 1500°C (2732°F). All of the tubing is of lamp-workable quality.