UVP High-Intensity UV Inspection Lamps, Analytik Jena
Supplier: Analytik Jena
95-0127-01 95-0127-03 95-0127-06 95-0127-02 95-0127-04 95-0127-07
36595-021EA 2972.15 CAD
36595-021 36595-022 36595-023 21474-638 21474-636 36595-020
UVP High-Intensity UV Inspection Lamps, Analytik Jena
Lights UV Lights, Lamps and Bulbs
Longwave 100 W lamps are designed for quality control, industrial inspection, and non-destructive testing. Lamp heads swivel 360° and the filter is recessed to prevent breakage. Cool-Touch* housing keeps lamp cool to the touch.
Model B-100YP features a 580 nm yellow filter which blocks wavelengths shorter than 500 nm and produces three strong mercury lines at 543, 574, and 576 nm
Models B-100AP and B-100AP/R provide a concentrated beam and high intensity of 21700 µW/cm2 at 5.1 cm (2") and 8900µW/cm2 at 25.4 cm (10") of 365 nm UV energy. Lamp head on Model B-100AP is connected to transformer by a 2.4 m (8') cord. Model B-100AP/R features 6.1 m (20') primary and secondary cords for added mobility. Lamp head dimensions: 24.8W×15.2 dia.cm (93/4×6").
Lamp head is connected to transformer by 2.4 m (8') cord.
Ordering information: Lamps include 100W spot bulb, transformer, and cord. Replaceable spot bulbs are sold separately. Optional lamp funnel (21474-508) and visor (21474-506) direct UV irradiance and shield operator’s eyes.