V-Maxx™ Goggles, Honeywell Safety
Supplier: Honeywell Safety Products
11250800 11250810
CA89045-006EA 8.97 CAD
CA89045-006 CA89045-008
V-Maxx™ Goggles, Honeywell Safety
Goggles
Sleek, wrap-around styling provides a clear and unobstructed 180° field of vision.
Goggles are equipped with headband clips that allow adjustment around hardhats or hearing protection, and they can be worn over most prescription eyewear. The clear lenses are coated with Fog-Ban® antifog treatment. Available in direct or indirect ventilation styles for impact or splash protection.
Certifications: Goggles meet ANSI Z87+ and CSA Z94.3 safety standards.