VACUETTE® Urine Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
455028 456007 455007
95057-267CS 1430.88 CAD
95057-267 95057-277 10809-036
VACUETTE® Urine Tubes, Greiner Bio-One
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Urine tubes are designed for use as collection, transportation, or analysis vessels.
  • Clear, durable plastic construction
  • Sterile
  • Leakproof and unbreakable
  • Disposable
Manufactured from clear, disposable plastic, these tubes are sterile, leak-proof, and virtually unbreakable
VACUETTE® urine tubes are also available with or without a stabilizing preservative. Tubes containing a preservative are intended for use with specimens that cannot be analyzed within two hours of collection, specimens being tested for an otherwise unstable analyte, or specimens being stabilized for microbiological studies. Tubes with preservative should be inverted 6–8 times following collection in order to acheive homogenous mixing.
Available in round or conical bottom formats. Round bottom tubes are used for chemical urine tests. Conical bottom tubes are used for microscopic examinations of urine sedimentation. All tubes are supplied with yellow caps.