Bard-Parker® Stainless Steel Scalpel Blades, Nos. 10–15, Sterile, Aspen Surgical
Supplier: Aspen Surgical Products
371210 371211 371212 371215
89176-362CS 284.84 CAD
89176-362 89176-364 89176-366 89176-368
Bard-Parker® Stainless Steel Scalpel Blades, Nos. 10–15, Sterile, Aspen Surgical
Scalpels and Scalpel Assemblies Scalpel Blades
Stainless steel blades are sterile and individually packaged
For use with No. 3, No. 3L, No. 3LA, No. 5, No. 7, and No. 9 Bard-Parker® nonsterile surgical handles.