VWR® Maximum Protection Covers, Impervious Fabric
Supplier: VWR
VWR®
414004-460
414004-460CS 336.35 CAD
414004-460
VWR® Maximum Protection Covers, Impervious Fabric
Covers
These covers are the cleanest and most durable within the entire VWR® Protection line. They provide barrier protection against fluids, biologicals, particulates, and light chemical splash.
  • Full protection from fluids and light chemical splash
  • Manufactured from fluid-impervious material
  • Lowest level of particle shedding
  • Clean, low-linting fabric
  • 99.9% bacterial filtration efficiency
Particle shedding Level I per Helmke Drum test.
Certifications: Covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each item meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.