WebSeal™ Standard Mats and Sealing Tapes, Non sterile
Supplier: Thermo Scientific
60180-M100 60180-M101 60180-M102 60180-M103 60180-M121 60180-M123 60180-M126 60180-M150 60180-M142 60180-M143 60180-M144 60180-M145 60180-M146 60180-M950 60180-M999 60180-M116 60180-M149 60180-M113 60180-M108 60180-M111 60180-M112 60180-M115 60180-M120 60180-M122 60180-M125 60180-M131 60180-M134 60180-M105 60180-M171 60180-M176 60180-M177 60180-M178 60180-M183 60180-M184 60180-M185 60180-M186 60180-M210 60180-M975 60180-M179
10036-174PK 340.41 CAD
10036-174 10036-176 10036-178 10036-180 10036-182 10036-184 10036-186 10036-188 10036-190 10036-192 10036-194 10036-196 10036-198 10036-200 10036-202 10036-234 10036-236 10036-238 10036-240 10036-242 10036-244 10036-246 10036-248 10036-250 10036-252 10036-254 75796-570 75800-264 75982-222 75982-224 75982-226 75982-228 75982-230 75982-232 75982-234 75982-236 75982-238 75982-240 76047-536
WebSeal™ Standard Mats and Sealing Tapes, Non sterile
Seals Microplate Seals
These sealing mats are placed directly on the microplate by hand or with an applicator.
Manufactured from pure silicone with wider or small penetration area for ease of use. Mats are dry heat autoclavable, provide excellent chemical compatibility, and resist coring and tearing. Pre-slit mats ensure easy penetration and reduce vacuum problems. Non sterile.