Barnstead Automatic Collection System, Thermo Scientific
Supplier: Thermo Fisher Scientific
440138 B440704 400634 440241
26299-454EA 448.58 CAD
26299-454 26299-385 26299-421 26299-487
Barnstead Automatic Collection System, Thermo Scientific
Water Purification Systems Stills
Designed to collect water from the Barnstead Mega-Pure™ stills and control the operation of the Barnstead Mega-Pure™ Automatic Systems.
For use with models MP-3A, MP-6A, and MP-11A (see series 26294-257)
Use the optional tubing adapter kit where wall space requires more compact system. The optional outlet adapter allows an automatic dishwasher to be connected to collection bottle outlet. Equipped with shut-off stopcock.
A fully self-tending system consisting of 45L PYREX® brand glass collection bottle, mounting platform with dispensing valve and automatic control unit. Built-inMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable