Flash Chromatography Columns (60 Å Pore Size), Agela Technologies
Supplier: Phenomenex
CS140800-0 CS140004-0 CS140012-0 CS140020-0 CS140040-0 CS140080-0 CS140120-0 CS140330-0 CM140004-0 CM140012-0 CM140040-0 CM140080-0 CM140020-0 CM140120-0 CS140220-0 CS1401500-0
89168-654EA 702.74 CAD
89168-654 97006-634 97006-636 97006-638 97006-640 97006-642 97006-644 97006-646 97009-734 97009-744 97009-800 97009-802 97009-816 97009-830 75807-864 76327-380
Flash Chromatography Columns (60 Å Pore Size), Agela Technologies
Chromatography Columns
Silica-based columns are made with high purity analytical grade silica and are suitable for most flash chromatography systems.
Special surface modification greatly reduces over-activity of the silica surface, which reduces tailing of compounds and ensures high recovery and good reproducibility. The unique HILIC columns allow the purification of highly polar compounds which are not suitable for bare silica or ODS columns.