VWR® Ladle, PTFE
Supplier: VWR
VWR®
037.010
89026-192EA 204.98 CAD
89026-192
VWR® Ladle, PTFE
Samplers Sample Dippers
This PTFE molded sampling ladle is useful for transferring hot or cold reactive substances.
  • Chemically resistant
  • Inert
  • PTFE fabrication