Synthware Solvent Distilling Head, Kemtech America
Supplier: Kemtech America
S251250 S254250 S254500 S254000
60004-774EA 585.48 CAD
60004-774 60004-778 60004-784 60004-786
Synthware Solvent Distilling Head, Kemtech America
Distillation Components Distillation Heads
The upper sampling port features a 4mm PTFE stopcock with a [ST] 14/20 outer joint for use with septa.
  • Heavy wall Borosilicate construction
  • Ground glass joints
  • Teflon® stopcocks
The lower drain is centered to allow total drainage of the storage head.